Assay Details
Target Gene Details
Entrez Gene ID: | 8604 |
Gene Name: | solute carrier family 25 member 12 |
Gene Aliases: |
AGC1, ARALAR |
Location: |
Chr.2:171783405-171894306 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A12 | NM_003705.4 | 18 | 2034 | NP_003696.2 |
| NR_047549.1 | 17 | 1918 | ||
| XM_011512070.2 | 19 | 2106 | XP_011510372.1 | |
| AJ496568.1 | 18 | 1963 | CAD43090.1 | |
| AK022828.1 | 18 | 1972 | ||
| AK054941.1 | 3 | 1351 | ||
| AK091071.1 | 17 | 1835 | ||
| BC016932.1 | 18 | 1951 | AAH16932.1 | |
| Y14494.1 | 18 | 1965 | CAA74834.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834457 | Chr.2:171708429 - 171914391 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map