Assay Details
Target Gene Details
Entrez Gene ID: | 638 |
Gene Name: | BCL2 interacting killer |
Gene Aliases: |
BIP1, BP4, NBK |
Location: |
Chr.22:43110748-43129712 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BIK | NM_001197.4 | NP_001188.1 | ||
| BC001599.1 | AAH01599.1 | |||
| BM765910.1 | ||||
| BP329261.1 | ||||
| BT006728.1 | AAP35374.1 | |||
| CR456390.1 | CAG30276.1 | |||
| CR541863.1 | CAG46661.1 | |||
| CR541883.1 | CAG46681.1 | |||
| U34584.1 | AAC50413.1 | |||
| U49730.1 | AAC79124.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762124 | Chr.22:43031130 - 43188670 on Build GRCh38 | Gain |
|
| esv33816 | Chr.22:43113758 - 43326122 on Build GRCh38 | Gain |
|
| nsv834214 | Chr.22:43037931 - 43217928 on Build GRCh38 | Gain |
|
| esv3647854 | Chr.22:43120675 - 43128505 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map