Assay Details
Target Gene Details
Entrez Gene ID: | 89874 |
Gene Name: | solute carrier family 25 member 21 |
Gene Aliases: |
ODC, ODC1 |
Location: |
Chr.14:36677921-37172660 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A21 | NM_001171170.1 | 1 | 295 | NP_001164641.1 |
| NM_030631.3 | 1 | 295 | NP_085134.1 | |
| XM_011537287.2 | XP_011535589.1 | |||
| AJ278148.1 | 1 | 295 | CAC27562.1 | |
| AK289575.1 | 1 | 58 | ||
| BC101521.1 | AAI01522.1 | |||
| BC113365.1 |
Target Gene Details
Entrez Gene ID: | 100129794 |
Gene Name: | SLC25A21 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.14:37172026-37173794 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A21-AS1 | NR_033240.1 | 1 | 341 | |
| AK057476.1 | 1 | 264 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1871n100 | Chr.14:37087152 - 37201854 on Build GRCh38 | Gain |
|
| nsv1153559 | Chr.14:37162398 - 37302029 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map