Assay Details
Target Gene Details
Entrez Gene ID: | 29071 |
Gene Name: | C1GALT1 specific chaperone 1 |
Gene Aliases: |
C1GALT2, C38H2-L1, COSMC, HSPC067, MST143, TNPS |
Location: |
Chr.X:120625674-120630150 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1GALT1C1 | NM_001011551.2 | 2 | 1174 | NP_001011551.1 |
| NM_152692.4 | 3 | 1346 | NP_689905.1 | |
| AB084170.1 | 2 | 1039 | BAC41493.1 | |
| AF088029.1 | 1 | 233 | ||
| AF161552.1 | 3 | 1188 | AAF29039.1 | |
| AF177284.1 | 2 | 1049 | AAQ13670.1 | |
| AJ238398.1 | 2 | 1172 | CAC80277.1 | |
| AK290111.1 | 2 | 1055 | ||
| AY159319.1 | 3 | 1191 | AAN78129.1 | |
| AY358642.1 | 2 | 1015 | AAQ89005.1 | |
| BC011930.2 | 2 | 993 | AAH11930.1 | |
| BC050441.1 | 2 | 1064 | AAH50441.1 | |
| BM971206.1 | 1 | 570 |
More Information
Set Membership: |
|

Genomic Map