Assay Details
Target Gene Details
Entrez Gene ID: | 79153 |
Gene Name: | glycerophosphodiester phosphodiesterase domain containing 3 |
Gene Aliases: |
GDE7 |
Location: |
Chr.16:30104810-30113557 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDPD3 | NM_024307.2 | NP_077283.2 | ||
| AK026256.1 | BAB15414.1 | |||
| AK225450.1 | ||||
| BC002714.2 | AAH02714.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833187 | Chr.16:30051939 - 30173160 on Build GRCh38 | Loss |
|
| nsv1160384 | Chr.16:30023148 - 30178660 on Build GRCh38 | Duplication |
|
| esv2758642 | Chr.16:30062634 - 30371959 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map