Assay Details
Target Gene Details
Entrez Gene ID: | 8649 |
Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 3 |
Gene Aliases: |
MAP2K1IP1, MAPBP, MAPKSP1, MP1, PRO0633, Ragulator3 |
Location: |
Chr.4:99878338-99894546 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMTOR3 | NM_001243736.1 | NP_001230665.1 | ||
| NM_021970.3 | NP_068805.1 | |||
| NR_024170.1 | ||||
| AF130115.1 | AAG35540.1 | |||
| AF201947.1 | AAF17239.1 | |||
| AK223311.1 | BAD97031.1 | |||
| AK311755.1 | ||||
| AL544981.3 | ||||
| BC026245.1 | AAH26245.1 | |||
| BI562630.1 |
More Information
Set Membership: |
|

Genomic Map