Assay Details
Target Gene Details
Entrez Gene ID: | 3716 |
Gene Name: | Janus kinase 1 |
Gene Aliases: |
JAK1A, JAK1B, JTK3 |
Location: |
Chr.1:64833223-65067746 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAK1 | NM_001320923.1 | NP_001307852.1 | ||
| NM_001321852.1 | NP_001308781.1 | |||
| NM_001321853.1 | NP_001308782.1 | |||
| NM_001321854.1 | NP_001308783.1 | |||
| NM_001321855.1 | NP_001308784.1 | |||
| NM_001321856.1 | NP_001308785.1 | |||
| NM_001321857.1 | NP_001308786.1 | |||
| NM_002227.3 | NP_002218.2 | |||
| XM_017001212.1 | XP_016856701.1 | |||
| AB209057.1 | BAD92294.1 | |||
| AB219242.1 | BAE02826.1 | |||
| AK314257.1 | ||||
| BC132729.1 | ||||
| BX647675.1 | ||||
| HQ628631.1 | ||||
| M64174.1 | AAA36527.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006347 | Chr.1:64839511 - 64850546 on Build GRCh38 | Gain+Loss |
|
| nsv1005169 | Chr.1:64838732 - 64850546 on Build GRCh38 | Gain |
|
| dgv172n100 | Chr.1:64838787 - 64869762 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map