Assay Details
Target Gene Details
Entrez Gene ID: | 9567 |
Gene Name: | GTP binding protein 1 |
Gene Aliases: |
GP-1, GP1, HSPC018 |
Location: |
Chr.22:38704561-38738265 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTPBP1 | NM_004286.4 | NP_004277.2 | ||
| XM_011530537.2 | 1 | 1260 | XP_011528839.1 | |
| XM_017029099.1 | 1 | 1252 | XP_016884588.1 | |
| XM_017029100.1 | 1 | 1246 | XP_016884589.1 | |
| BF001482.1 |
More Information
Set Membership: |
|

Genomic Map