Assay Details
Target Gene Details
Entrez Gene ID: | 23404 |
Gene Name: | exosome component 2 |
Gene Aliases: |
RRP4, Rrp4p, hRrp4p, p7 |
Location: |
Chr.9:130693760-130704894 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOSC2 | NM_001282708.1 | NP_001269637.1 | ||
| NM_001282709.1 | NP_001269638.1 | |||
| NM_014285.6 | NP_055100.2 | |||
| NR_104230.1 | ||||
| XM_005272176.2 | XP_005272233.1 | |||
| XM_006717023.2 | XP_006717086.1 | |||
| XM_017014558.1 | 3 | 301 | XP_016870047.1 | |
| AK001916.1 | BAA91977.1 | |||
| AK022460.1 | BAB14043.1 | |||
| AK090512.1 | 3 | 301 | ||
| AK296605.1 | ||||
| AK303163.1 | ||||
| BC000747.2 | AAH00747.1 | |||
| DA813307.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621866 | Chr.9:130695968 - 131205700 on Build GRCh38 | Gain |
|
| nsv825122 | Chr.9:130637156 - 130697917 on Build GRCh38 | Gain |
|
| nsv831735 | Chr.9:130598942 - 130791581 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map