Assay Details
Target Gene Details
Entrez Gene ID: | 64965 |
Gene Name: | mitochondrial ribosomal protein S9 |
Gene Aliases: |
MRP-S9, RPMS9, S9mt |
Location: |
Chr.2:105038025-105099960 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS9 | NM_182640.2 | NP_872578.1 | ||
| XM_011511644.2 | XP_011509946.1 | |||
| AK001451.1 | ||||
| AK130078.1 | ||||
| BC047784.1 | AAH47784.1 | |||
| BC057240.1 | AAH57240.1 |
Target Gene Details
Entrez Gene ID: | 101927492 |
Gene Name: | uncharacterized LOC101927492 |
Gene Aliases: |
- |
Location: |
Chr.2:105097052-105102944 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UTAT33 | NR_110227.1 | |||
| CR736606.1 |
More Information
Set Membership: |
|

Genomic Map