Assay Details
Target Gene Details
Entrez Gene ID: | 8996 |
Gene Name: | nucleolar protein 3 |
Gene Aliases: |
ARC, FCM, MYP, NOP, NOP30 |
Location: |
Chr.16:67170497-67175737 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOL3 | NM_001185057.2 | 4 | 1068 | NP_001171986.1 |
| NM_001276307.1 | 6 | 1102 | NP_001263236.1 | |
| NM_001276309.1 | 4 | 962 | NP_001263238.1 | |
| NM_001276311.1 | 4 | 954 | NP_001263240.1 | |
| NM_001276312.1 | 6 | 1453 | NP_001263241.1 | |
| NM_001276319.1 | 6 | 1453 | NP_001263248.1 | |
| NM_003946.6 | 4 | 1078 | NP_003937.1 | |
| XM_005256217.2 | 4 | 1705 | XP_005256274.1 | |
| XM_005256219.3 | 4 | 1591 | XP_005256276.1 | |
| XM_011523424.2 | 5 | 1550 | XP_011521726.1 | |
| XM_017023843.1 | 4 | 1715 | XP_016879332.1 | |
| AF064599.1 | 4 | 919 | AAC18593.1 | |
| AK294145.1 | 6 | 1418 | ||
| BC012798.2 | 4 | 964 | AAH12798.1 | |
| BP215200.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion |
|
| nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss |
|
| nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
| nsv471092 | Chr.16:67174030 - 67518392 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map