Assay Details
Target Gene Details
Entrez Gene ID: | 2804 |
Gene Name: | golgin B1 |
Gene Aliases: |
GCP, GCP372, GOLIM1 |
Location: |
Chr.3:121663199-121749908 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLGB1 | NM_001256486.1 | 20 | 9658 | NP_001243415.1 |
| NM_001256487.1 | 20 | 9671 | NP_001243416.1 | |
| NM_001256488.1 | 19 | 9548 | NP_001243417.1 | |
| NM_004487.4 | 20 | 9628 | NP_004478.3 | |
| XM_005247371.4 | 20 | 10118 | XP_005247428.1 | |
| XM_005247372.4 | 20 | 9694 | XP_005247429.1 | |
| XM_005247373.1 | 19 | 9510 | XP_005247430.1 | |
| XM_006713587.1 | 20 | 9618 | XP_006713650.1 | |
| XM_006713588.1 | 20 | 9618 | XP_006713651.1 | |
| XM_006713589.1 | 20 | 9615 | XP_006713652.1 | |
| XM_006713590.1 | 19 | 9495 | XP_006713653.1 | |
| XM_006713591.1 | 18 | 9393 | XP_006713654.1 | |
| XM_011512699.2 | 20 | 9585 | XP_011511001.1 | |
| XM_017006189.1 | 20 | 10103 | XP_016861678.1 | |
| XM_017006190.1 | 20 | 10270 | XP_016861679.1 | |
| XM_017006191.1 | 19 | 9995 | XP_016861680.1 | |
| XM_017006192.1 | 20 | 9679 | XP_016861681.1 | |
| XM_017006193.1 | 20 | 9676 | XP_016861682.1 | |
| XM_017006194.1 | 19 | 9571 | XP_016861683.1 | |
| XM_017006195.1 | 14 | 9540 | XP_016861684.1 | |
| AB371588.1 | 20 | 9631 | ||
| AB593126.1 | 19 | 9522 | ||
| D25542.1 | 20 | 9626 | BAA05025.1 | |
| X75304.1 | 20 | 9616 | CAA53052.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8524n54 | Chr.3:121252053 - 121838686 on Build GRCh38 | Gain |
|
| esv3575599 | Chr.3:121571453 - 121671424 on Build GRCh38 | Gain |
|
| nsv829703 | Chr.3:121664443 - 121815646 on Build GRCh38 | Loss |
|
| esv3575598 | Chr.3:121406926 - 121715587 on Build GRCh38 | Gain |
|
| nsv998740 | Chr.3:121570652 - 121671424 on Build GRCh38 | Gain |
|
| nsv999335 | Chr.3:121381436 - 121689919 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map