Assay Details
Target Gene Details
Entrez Gene ID: | 5892 |
Gene Name: | RAD51 paralog D |
Gene Aliases: |
BROVCA4, R51H3, RAD51L3, TRAD |
Location: |
Chr.17:35099792-35119869 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD51D | NM_001142571.1 | NP_001136043.1 | ||
| NM_002878.3 | NP_002869.3 | |||
| NM_133629.2 | NP_598332.1 | |||
| NR_037711.1 | ||||
| NR_037712.1 | ||||
| AB013341.1 | BAA25914.1 | |||
| AB016223.1 | BAA31747.1 | |||
| AB016224.1 | BAA31748.1 | |||
| AB016225.1 | BAA31749.1 | |||
| AB020412.1 | BAA34690.1 | |||
| AF034956.1 | AAC39719.1 | |||
| AK296241.1 | ||||
| AK301249.1 | ||||
| AL117459.1 | CAB55937.1 | |||
| BC002723.2 | AAH02723.1 | |||
| BC014422.1 | AAH14422.1 | |||
| BX647297.1 | ||||
| Y15572.1 | CAA75681.1 |
Target Gene Details
Entrez Gene ID: | 100529207 |
Gene Name: | RAD51L3-RFFL readthrough |
Gene Aliases: |
- |
Location: |
Chr.17:35009112-35121522 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD51L3-RFFL | NR_037714.1 | |||
| AK097811.1 |
More Information
Set Membership: |
|

Genomic Map