Assay Details
Target Gene Details
Entrez Gene ID: | 6472 |
Gene Name: | serine hydroxymethyltransferase 2 |
Gene Aliases: |
GLYA, HEL-S-51e, SHMT |
Location: |
Chr.12:57229573-57234935 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHMT2 | NM_001166356.1 | 12 | 1828 | NP_001159828.1 |
| NM_001166357.1 | 12 | 1920 | NP_001159829.1 | |
| NM_001166358.1 | 12 | 1819 | NP_001159830.1 | |
| NM_001166359.1 | 12 | 1712 | NP_001159831.1 | |
| NM_005412.5 | 12 | 1858 | NP_005403.2 | |
| NR_029415.1 | 11 | 1660 | ||
| NR_029416.1 | 11 | 1739 | ||
| NR_029417.1 | 11 | 1778 | ||
| NR_048562.1 | 11 | 1868 | ||
| XM_011538675.2 | 12 | 1798 | XP_011536977.1 | |
| XM_011538676.2 | 12 | 1789 | XP_011536978.1 | |
| XM_011538677.2 | 11 | 1699 | XP_011536979.1 | |
| XM_011538678.2 | 11 | 1681 | XP_011536980.1 | |
| AK055053.1 | 6 | 3184 | ||
| AK223555.1 | 12 | 1720 | BAD97275.1 | |
| AK295941.1 | 11 | 1714 | ||
| AK296183.1 | ||||
| AK297173.1 | 11 | 1620 | ||
| AK301339.1 | ||||
| AK301443.1 | ||||
| AK303827.1 | 11 | 1534 | ||
| AK315916.1 | 12 | 1912 | ||
| BC011911.2 | 12 | 1662 | AAH11911.1 | |
| BC013677.2 | 12 | 1722 | AAH13677.1 | |
| BC032584.1 | 12 | 1701 | AAH32584.1 | |
| BC044211.1 | 12 | 1698 | AAH44211.1 | |
| BC091501.1 | 11 | 1582 | AAH91501.1 | |
| BX647711.1 | 12 | 1779 | CAI46021.1 | |
| BX648847.1 | 10 | 1260 | ||
| EU668344.1 | 12 | 1662 | ||
| L11932.1 | 11 | 1563 | AAA63258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| nsv952189 | Chr.12:57127118 - 57245617 on Build GRCh38 | Deletion |
|
| nsv519293 | Chr.12:57189889 - 57243810 on Build GRCh38 | Loss |
|
| dgv96n21 | Chr.12:57139199 - 57243810 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map