Assay Details
Target Gene Details
Entrez Gene ID: | 90268 |
Gene Name: | OTU deubiquitinase with linear linkage specificity |
Gene Aliases: |
FAM105B, GUM |
Location: |
Chr.5:14660794-14716552 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OTULIN | NM_138348.4 | NP_612357.4 | ||
| XM_011514151.2 | XP_011512453.1 | |||
| XM_011514152.2 | XP_011512454.1 | |||
| XM_011514154.2 | XP_011512456.1 | |||
| XM_017010015.1 | XP_016865504.1 | |||
| BC015392.1 | AAH15392.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026914 | Chr.5:14237673 - 14751479 on Build GRCh38 | Gain |
|
| nsv830211 | Chr.5:14474566 - 14675667 on Build GRCh38 | Loss |
|
| nsv1018187 | Chr.5:14385055 - 15126994 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map