Assay Details
Target Gene Details
Entrez Gene ID: | 6197 |
Gene Name: | ribosomal protein S6 kinase A3 |
Gene Aliases: |
CLS, HU-3, ISPK-1, MAPKAPK1B, MRX19, RSK, RSK2, S6K-alpha3, p90-RSK2, pp90RSK2 |
Location: |
Chr.X:20149911-20267514 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526033 | Chr.X:20149203 - 20206450 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map