Assay Details
Target Gene Details
Entrez Gene ID: | 89944 |
Gene Name: | galactosidase beta 1 like 2 |
Gene Aliases: |
MST114, MSTP114 |
Location: |
Chr.11:134331874-134376324 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLB1L2 | NM_138342.3 | NP_612351.2 | ||
| XM_006718937.1 | XP_006719000.1 | |||
| AF173889.1 | AAQ13636.1 | |||
| AK074703.1 | BAC11149.1 | |||
| AK074855.1 | BAC11247.1 | |||
| AK074947.1 | BAC11309.1 | |||
| AY358497.1 | AAQ88861.1 | |||
| BC040641.1 | AAH40641.1 | |||
| BX640934.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1302n100 | Chr.11:134275705 - 134348093 on Build GRCh38 | Loss |
|
| dgv4e196 | Chr.11:134304439 - 134744665 on Build GRCh38 | Duplication |
|
| nsv521581 | Chr.11:134126584 - 134772738 on Build GRCh38 | Gain |
|
| dgv1304n100 | Chr.11:134322594 - 134754120 on Build GRCh38 | Gain |
|
| esv2760555 | Chr.11:134282136 - 134860776 on Build GRCh38 | Gain+Loss |
|
| nsv521623 | Chr.11:134307381 - 134756993 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map