Assay Details
Target Gene Details
Entrez Gene ID: | 10446 |
Gene Name: | leucine rich repeat neuronal 2 |
Gene Aliases: |
FIGLER7, GAC1, LRANK1, LRRN5 |
Location: |
Chr.1:204617170-204685836 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRN2 | NM_006338.2 | 3 | 632 | NP_006329.2 |
| NM_201630.1 | 2 | 620 | NP_963924.1 | |
| XM_005244827.3 | 3 | 1090 | XP_005244884.1 | |
| XM_011509066.2 | 3 | 702 | XP_011507368.1 | |
| XM_017000050.1 | 2 | 2144 | XP_016855539.1 | |
| AF030435.1 | 3 | 632 | AAC39792.1 | |
| AK226055.1 | 2 | 509 | ||
| AK312408.1 | 3 | 625 | ||
| AL833565.1 | 1 | 2457 | ||
| AY358290.1 | 1 | 1124 | AAQ88657.1 | |
| BC034047.1 | 2 | 620 | AAH34047.1 | |
| BC068541.1 | 2 | 620 | AAH68541.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4154 | Chr.1:204556130 - 204620074 on Build GRCh38 | Insertion |
|
| esv3588622 | Chr.1:204585680 - 204625853 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map