Assay Details
Target Gene Details
Entrez Gene ID: | 23286 |
Gene Name: | WW and C2 domain containing 1 |
Gene Aliases: |
HBEBP3, HBEBP36, KIBRA, MEMRYQTL, PPP1R168 |
Location: |
Chr.5:168292060-168472303 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWC1 | NM_001161661.1 | NP_001155133.1 | ||
| NM_001161662.1 | NP_001155134.1 | |||
| NM_015238.2 | NP_056053.1 | |||
| XM_005265850.1 | XP_005265907.1 | |||
| XM_005265853.2 | XP_005265910.1 | |||
| XM_011534485.1 | XP_011532787.1 | |||
| XM_011534486.1 | XP_011532788.1 | |||
| XM_011534487.1 | XP_011532789.1 | |||
| XM_011534488.1 | XP_011532790.1 | |||
| XM_011534489.1 | XP_011532791.1 | |||
| XM_011534490.1 | XP_011532792.1 | |||
| XM_011534491.1 | XP_011532793.1 | |||
| XM_017009276.1 | XP_016864765.1 | |||
| AF506799.1 | AAO15881.1 | |||
| AK296323.1 | ||||
| BX640827.1 | CAE45903.1 | |||
| DR001014.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1074343 | Chr.5:168291694 - 168292895 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map