Assay Details
Target Gene Details
Entrez Gene ID: | 64376 |
Gene Name: | IKAROS family zinc finger 5 |
Gene Aliases: |
PEGASUS, ZNFN1A5 |
Location: |
Chr.10:122990806-123008850 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IKZF5 | NM_001271840.1 | 5 | 1432 | NP_001258769.1 |
| NR_073475.1 | 5 | 1476 | ||
| NR_073476.1 | 5 | 1472 | ||
| NR_073477.1 | 5 | 1463 | ||
| NR_073478.1 | 4 | 1325 | ||
| NR_073479.1 | 6 | 1584 | ||
| XM_006717947.3 | 6 | 1546 | XP_006718010.1 | |
| XM_017016550.1 | 5 | 1424 | XP_016872039.1 | |
| XM_017016551.1 | 6 | 1520 | XP_016872040.1 | |
| XM_017016552.1 | 5 | 1412 | XP_016872041.1 | |
| XM_017016553.1 | 5 | 1921 | XP_016872042.1 | |
| XM_017016554.1 | 6 | 2029 | XP_016872043.1 | |
| XM_017016555.1 | 5 | 1908 | XP_016872044.1 | |
| AF230808.1 | 3 | 1122 | AAG39220.1 | |
| AK026626.1 | 5 | 1421 | BAB15512.1 | |
| AK122899.1 | 6 | 3223 | ||
| AK226143.1 | 5 | 1413 | ||
| BX647958.1 | 5 | 1425 | CAH56199.1 | |
| CR749800.1 | 5 | 1418 | CAH18660.1 |
Target Gene Details
Entrez Gene ID: | 118672 |
Gene Name: | phosphoseryl-tRNA kinase |
Gene Aliases: |
C10orf89 |
Location: |
Chr.10:122980040-122997519 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSTK | AK094271.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7586 | Chr.10:122982687 - 123016052 on Build GRCh38 | Insertion |
|
| nsv832010 | Chr.10:122826964 - 123037874 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map