Assay Details
Target Gene Details
Entrez Gene ID: | 56852 |
Gene Name: | RAD18, E3 ubiquitin protein ligase |
Gene Aliases: |
RNF73 |
Location: |
Chr.3:8877196-8969249 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD18 | NM_020165.3 | 13 | 1586 | NP_064550.3 |
| XM_017006873.1 | 14 | 1744 | XP_016862362.1 | |
| AB035274.2 | 13 | 1621 | BAA99284.1 | |
| AF169796.1 | 13 | 1566 | AAF80856.1 | |
| AK023075.1 | 13 | 1573 | BAB14392.1 | |
| AK222771.1 | BAD96491.1 | |||
| AY004333.1 | 13 | 1532 | AAF86618.1 | |
| BC001302.1 | 13 | 1553 | AAH01302.1 |
More Information
Set Membership: |
|

Genomic Map