Assay Details
Target Gene Details
Entrez Gene ID: | 9048 |
Gene Name: | artemin |
Gene Aliases: |
ENOVIN, EVN, NBN |
Location: |
Chr.1:43933320-43937240 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARTN | NM_001136215.1 | 1 | 218 | NP_001129687.1 |
| NM_057090.2 | 1 | 218 | NP_476431.2 | |
| NM_057091.2 | 1 | 218 | NP_476432.2 | |
| AF120274.1 | 1 | 218 | AAD21075.1 |
More Information
Set Membership: |
|

Genomic Map