Assay Details
Target Gene Details
Entrez Gene ID: | 5269 |
Gene Name: | serpin family B member 6 |
Gene Aliases: |
CAP, DFNB91, MSTP057, PI-6, PI6, PTI, SPI3 |
Location: |
Chr.6:2948159-2972165 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB6 | NM_001195291.2 | NP_001182220.2 | ||
| NM_001271824.1 | NP_001258753.1 | |||
| NM_001297699.1 | 1 | 644 | NP_001284628.1 | |
| NM_001297700.1 | NP_001284629.1 | |||
| NM_004568.5 | NP_004559.4 | |||
| XM_011514672.1 | XP_011512974.1 | |||
| XM_017010940.1 | XP_016866429.1 | |||
| AK094983.1 | ||||
| AK314578.1 | ||||
| BC098564.1 | AAH98564.1 | |||
| BG499326.1 | ||||
| BI259189.1 | ||||
| BI827451.1 | ||||
| BM789272.1 | ||||
| BP325314.1 | ||||
| BX375950.2 | 1 | 84 | ||
| CA847860.1 | ||||
| CD674931.1 | ||||
| Z22658.1 | CAA80373.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600833 | Chr.6:2970334 - 3010915 on Build GRCh38 | Loss |
|
| nsv1145088 | Chr.6:2971066 - 2971566 on Build GRCh38 | Deletion |
|
| nsv830575 | Chr.6:2847376 - 3035682 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map