Assay Details
Target Gene Details
Entrez Gene ID: | 79869 |
Gene Name: | cleavage and polyadenylation specific factor 7 |
Gene Aliases: |
CFIm59 |
Location: |
Chr.11:61402648-61429992 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPSF7 | NM_001136040.2 | 6 | 773 | NP_001129512.1 |
| NM_001142565.1 | 6 | 746 | NP_001136037.1 | |
| NM_024811.3 | 6 | 849 | NP_079087.3 | |
| XM_005274298.4 | 6 | 832 | XP_005274355.1 | |
| XM_005274299.4 | 6 | 776 | XP_005274356.1 | |
| XM_005274303.4 | 7 | 804 | XP_005274360.1 | |
| XM_011545257.2 | 6 | 859 | XP_011543559.1 | |
| XM_011545258.2 | 6 | 730 | XP_011543560.1 | |
| XM_011545259.2 | 6 | 703 | XP_011543561.1 | |
| XM_011545260.2 | 7 | 850 | XP_011543562.1 | |
| XM_011545261.2 | 7 | 851 | XP_011543563.1 | |
| XM_011545262.2 | 7 | 797 | XP_011543564.1 | |
| XM_011545263.2 | 7 | 796 | XP_011543565.1 | |
| XM_017018345.1 | 6 | 832 | XP_016873834.1 | |
| AJ275970.1 | 5 | 639 | CAC81661.1 | |
| AK022591.1 | 1 | 41 | BAB14118.1 | |
| AK092037.1 | ||||
| AK096343.1 | 6 | 729 | ||
| AK294578.1 | 6 | 702 | ||
| AL832546.1 | 4 | 444 | ||
| AL832667.1 | 6 | 737 | ||
| AL833332.1 | 6 | 702 | ||
| BC018135.1 | 6 | 739 | AAH18135.1 | |
| BX537888.1 | 6 | 727 | CAD97884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832178 | Chr.11:61402074 - 61573390 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map