Assay Details
Target Gene Details
Entrez Gene ID: | 222537 |
Gene Name: | heparan sulfate-glucosamine 3-sulfotransferase 5 |
Gene Aliases: |
3-OST-5, 3OST5, HS3OST5, NBLA04021 |
Location: |
Chr.6:114055586-114342388 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HS3ST5 | NM_153612.3 | NP_705840.2 | ||
| XM_006715379.2 | XP_006715442.1 | |||
| XM_011535588.2 | XP_011533890.1 | |||
| XM_017010470.1 | XP_016865959.1 | |||
| XM_017010471.1 | XP_016865960.1 | |||
| XM_017010472.1 | XP_016865961.1 | |||
| XM_017010473.1 | XP_016865962.1 | |||
| XM_017010474.1 | XP_016865963.1 | |||
| AF503292.1 | AAN37737.1 | |||
| AK126930.1 | ||||
| AK289907.1 | ||||
| BC093911.1 | AAH93911.1 | |||
| BC093913.1 | AAH93913.1 |
Target Gene Details
Entrez Gene ID: | 101927768 |
Gene Name: | uncharacterized LOC101927768 |
Gene Aliases: |
- |
Location: |
Chr.6:113969701-114340729 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927768 | NR_125845.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs148758563] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map