Assay Details
Target Gene Details
Entrez Gene ID: | 84873 |
Gene Name: | adhesion G protein-coupled receptor G7 |
Gene Aliases: |
GPR128 |
Location: |
Chr.3:100609589-100696174 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRG7 | NM_032787.2 | NP_116176.2 | ||
| XM_011513245.2 | XP_011511547.1 | |||
| AK027360.1 | BAB55061.1 | |||
| AY181246.1 | AAO27358.1 | |||
| BC113451.1 | ||||
| BC130388.1 | ||||
| BP270648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516418 | Chr.3:100605598 - 100714748 on Build GRCh38 | Gain |
|
| dgv613n27 | Chr.3:100591968 - 100714748 on Build GRCh38 | Gain |
|
| dgv4830n100 | Chr.3:100605431 - 100729339 on Build GRCh38 | Gain |
|
| dgv182e55 | Chr.3:100580466 - 100760540 on Build GRCh38 | Gain |
|
| nsv822201 | Chr.3:100606475 - 100610716 on Build GRCh38 | Loss |
|
| esv2675173 | Chr.3:100605143 - 100610867 on Build GRCh38 | Deletion |
|
| dgv1397e212 | Chr.3:100607666 - 100723653 on Build GRCh38 | Gain |
|
| dgv917e214 | Chr.3:100604090 - 100730204 on Build GRCh38 | Gain |
|
| esv3597176 | Chr.3:100605159 - 100610785 on Build GRCh38 | Loss |
|
| esv2763708 | Chr.3:100599084 - 100723665 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map