Assay Details
Target Gene Details
Entrez Gene ID: | 4901 |
Gene Name: | neural retina leucine zipper |
Gene Aliases: |
D14S46E, NRL-MAF, RP27 |
Location: |
Chr.14:24078693-24114924 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRL | XM_005267708.4 | XP_005267765.1 | ||
| XM_005267710.3 | XP_005267767.1 | |||
| XM_017021350.1 | XP_016876839.1 | |||
| BX161381.1 | CAD61873.1 |
Target Gene Details
Entrez Gene ID: | 5106 |
Gene Name: | phosphoenolpyruvate carboxykinase 2, mitochondrial |
Gene Aliases: |
PEPCK, PEPCK-M, PEPCK2 |
Location: |
Chr.14:24094131-24104132 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCK2 | NM_001018073.2 | 2 | 415 | NP_001018083.2 |
| NM_001291556.1 | NP_001278485.1 | |||
| NM_001308054.1 | 2 | 676 | NP_001294983.1 | |
| NM_004563.3 | 2 | 415 | NP_004554.3 | |
| XM_006720158.2 | 2 | 408 | XP_006720221.1 | |
| AK129934.1 | 2 | 233 | ||
| AK301400.1 | 2 | 259 | ||
| AK307913.1 | 2 | 644 | ||
| AK311710.1 | 2 | 646 | ||
| AK316206.1 | ||||
| BC001454.2 | 2 | 205 | AAH01454.1 | |
| BQ957219.1 | 2 | 633 | ||
| BX248272.1 | 2 | 189 | CAD62600.1 | |
| CR456913.1 | 2 | 140 | CAG33194.1 | |
| DB028389.1 | 2 | 408 | ||
| HY022221.1 | 3 | 315 | ||
| KU178243.1 | 2 | 140 | ||
| KU178244.1 | 2 | 140 | ||
| KU178245.1 | ||||
| X92720.1 | 2 | 206 | CAA63380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564057 | Chr.14:24056425 - 24149650 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map