Assay Details
Target Gene Details
Entrez Gene ID: | 115352 |
Gene Name: | Fc receptor like 3 |
Gene Aliases: |
CD307c, FCRH3, IFGP3, IRTA3, SPAP2 |
Location: |
Chr.1:157676481-157700985 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCRL3 | NM_001320333.1 | NP_001307262.1 | ||
| NM_052939.3 | NP_443171.2 | |||
| NR_135214.1 | ||||
| NR_135215.1 | ||||
| NR_135216.1 | ||||
| NR_135217.1 | ||||
| XM_006711145.1 | XP_006711208.1 | |||
| AF416901.1 | AAL13290.1 | |||
| AF416902.1 | AAL13291.1 | |||
| AF416903.1 | AAL13292.1 | |||
| AF416904.1 | AAL13293.1 | |||
| AF416905.1 | AAL13294.1 | |||
| AF459027.1 | AAL59390.1 | |||
| AK098122.1 | ||||
| AK301257.1 | ||||
| AL833391.1 | ||||
| AY043466.1 | AAK91779.1 | |||
| BC028933.1 | AAH28933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761949 | Chr.1:157626296 - 157696854 on Build GRCh38 | Loss |
|
| nsv520011 | Chr.1:157678308 - 157703838 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map