Assay Details
Target Gene Details
Entrez Gene ID: | 2595 |
Gene Name: | glucosidase alpha, neutral C |
Gene Aliases: |
- |
Location: |
Chr.15:42273658-42353666 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GANC | NM_001301409.1 | 6 | 1101 | NP_001288338.1 |
| NM_198141.2 | 5 | 714 | NP_937784.2 | |
| AF525397.1 | AAO14993.1 | |||
| AF545045.1 | 5 | 408 | AAN74756.1 | |
| AF545046.1 | 5 | 408 | AAN74757.1 | |
| AK074037.1 | 5 | 606 | ||
| AK074330.1 | 6 | 1103 | ||
| BC059406.1 | 5 | 586 | AAH59406.1 | |
| BC093833.1 | 5 | 431 | AAH93833.1 | |
| BC112051.1 | 5 | 431 | AAI12052.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636263 | Chr.15:42288763 - 42328316 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112976629] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map