Assay Details
Target Gene Details
Entrez Gene ID: | 84691 |
Gene Name: | family with sequence similarity 71 member F1 |
Gene Aliases: |
FAM137A, NYD-SP18 |
Location: |
Chr.7:128709061-128731743 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM71F1 | NM_001282788.1 | 7 | 1690 | NP_001269717.1 |
| NM_001282789.1 | 8 | 1637 | NP_001269718.1 | |
| NM_032599.3 | 7 | 1696 | NP_115988.1 | |
| NR_104242.1 | 8 | 1994 | ||
| NR_104243.1 | 9 | 1883 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2670312 | Chr.7:128499519 - 128999613 on Build GRCh38 | Deletion |
|
| esv2735137 | Chr.7:128499440 - 128999664 on Build GRCh38 | Deletion |
|

Genomic Map