Assay Details
Target Gene Details
Entrez Gene ID: | 10544 |
Gene Name: | protein C receptor |
Gene Aliases: |
CCCA, CCD41, EPCR |
Location: |
Chr.20:35171937-35215989 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROCR | NM_006404.4 | 4 | 1255 | NP_006395.2 |
| XM_011528496.1 | XP_011526798.1 | |||
| BC014451.1 | 4 | 1119 | AAH14451.1 | |
| BQ939781.1 | 2 | 587 | ||
| L35545.1 | 4 | 1061 | AAA63647.1 |
More Information
Set Membership: |
|

Genomic Map