Assay Details
Target Gene Details
Entrez Gene ID: | 25959 |
Gene Name: | KN motif and ankyrin repeat domains 2 |
Gene Aliases: |
ANKRD25, MXRA3, PPKWH, SIP |
Location: |
Chr.19:11164267-11197567 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KANK2 | NM_001136191.2 | 13 | 4591 | NP_001129663.1 |
| NM_015493.6 | 11 | 4333 | NP_056308.3 | |
| AB040951.2 | 12 | 4543 | ||
| AB284125.1 | 14 | 4774 | ||
| AL117489.1 | 1 | 1347 | CAB55958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065030 | Chr.19:11149372 - 11199804 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map