Assay Details
Target Gene Details
Entrez Gene ID: | 57628 |
Gene Name: | dipeptidyl peptidase like 10 |
Gene Aliases: |
DPL2, DPPY, DPRP-3, DPRP3 |
Location: |
Chr.2:114442358-115845752 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPP10 | NM_001321907.1 | 1 | 435 | NP_001308836.1 |
| NM_001321910.1 | 1 | 435 | NP_001308839.1 | |
| NM_001321913.1 | 1 | 435 | NP_001308842.1 | |
| NM_020868.4 | 1 | 435 | NP_065919.2 | |
| AK289967.1 | 1 | 134 | ||
| AK315147.1 | 1 | 237 | ||
| AY172661.3 | AAO17263.2 | |||
| BC030832.1 | 1 | 471 | AAH30832.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834337 | Chr.2:114412237 - 114584296 on Build GRCh38 | Gain |
|
| esv3592044 | Chr.2:114397692 - 114726043 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map