Assay Details
Target Gene Details
Entrez Gene ID: | 6541 |
Gene Name: | solute carrier family 7 member 1 |
Gene Aliases: |
ATRC1, CAT-1, ERR, HCAT1, REC1L |
Location: |
Chr.13:29509410-29595715 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A1 | NM_003045.4 | 13 | 3932 | NP_003036.1 |
| XM_005266507.3 | 13 | 3727 | XP_005266564.1 | |
| XM_017020713.1 | 13 | 3999 | XP_016876202.1 | |
| XM_017020714.1 | 13 | 3938 | XP_016876203.1 | |
| XM_017020715.1 | 13 | 3820 | XP_016876204.1 | |
| XM_017020716.1 | 13 | 3943 | XP_016876205.1 | |
| AB105377.1 | 13 | 3819 | ||
| AK124591.1 | 1 | 1254 | ||
| BC063303.1 | 13 | 3729 | AAH63303.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv561392 | Chr.13:29497108 - 29517682 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map