Assay Details
Target Gene Details
Entrez Gene ID: | 11052 |
Gene Name: | cleavage and polyadenylation specific factor 6 |
Gene Aliases: |
CFIM, CFIM68, HPBRII-4, HPBRII-7 |
Location: |
Chr.12:69239537-69274358 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPSF6 | NM_001300947.1 | 7 | 991 | NP_001287876.1 |
| NM_007007.2 | 6 | 880 | NP_008938.2 | |
| XM_005268588.2 | 7 | 991 | XP_005268645.1 | |
| XM_005268590.2 | 6 | 880 | XP_005268647.1 | |
| AK223568.1 | 6 | 845 | BAD97288.1 | |
| AK292024.1 | 6 | 778 | ||
| AK299928.1 | ||||
| BC000714.2 | 7 | 912 | AAH00714.1 | |
| BC005000.1 | AAH05000.1 | |||
| X67337.1 | 6 | 804 | CAA47752.1 |
More Information
Set Membership: |
|

Genomic Map