Assay Details
Target Gene Details
Entrez Gene ID: | 29108 |
Gene Name: | PYD and CARD domain containing |
Gene Aliases: |
ASC, CARD5, TMS, TMS-1, TMS1 |
Location: |
Chr.16:31201486-31202776 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYCARD | NM_013258.4 | 3 | 512 | NP_037390.2 |
| NM_145182.2 | 2 | 455 | NP_660183.1 | |
| AB023416.2 | 3 | 512 | BAA87339.2 | |
| AF184073.1 | 3 | 500 | AAG01188.1 | |
| AF255794.1 | 2 | 443 | AAF99665.1 | |
| AF310103.1 | 3 | 426 | AAG30286.1 | |
| AF384665.1 | 3 | 479 | AAK63850.1 | |
| AK000211.1 | 3 | 464 | BAA91012.1 | |
| AK314422.1 | 3 | 504 | ||
| BC004470.2 | 4 | 482 | AAH04470.1 | |
| BC013569.1 | 3 | 419 | AAH13569.2 |
More Information
Set Membership: |
|

Genomic Map