Assay Details
Target Gene Details
Entrez Gene ID: | 9826 |
Gene Name: | Rho guanine nucleotide exchange factor 11 |
Gene Aliases: |
GTRAP48, PDZ-RHOGEF |
Location: |
Chr.1:156934840-157046903 on Build GRCh38 |
Assay Gene Location: | Within Exon 47 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF11 | NM_014784.3 | 40 | 5782 | NP_055599.1 |
| NM_198236.2 | 41 | 5902 | NP_937879.1 | |
| XM_005245629.4 | 41 | 6306 | XP_005245686.1 | |
| XM_005245633.4 | 31 | 4165 | XP_005245690.1 | |
| XM_006711659.2 | 43 | 6354 | XP_006711722.1 | |
| XM_006711660.3 | 43 | 6351 | XP_006711723.1 | |
| XM_006711661.2 | 42 | 6324 | XP_006711724.1 | |
| XM_006711662.2 | 42 | 6321 | XP_006711725.1 | |
| XM_006711663.3 | 41 | 5341 | XP_006711726.1 | |
| XM_006711665.3 | 42 | 6231 | XP_006711728.1 | |
| XM_011510185.2 | 42 | 6339 | XP_011508487.1 | |
| XM_011510186.2 | 42 | 6234 | XP_011508488.1 | |
| XM_011510187.2 | 42 | 5065 | XP_011508489.1 | |
| XM_011510189.2 | 20 | 4370 | XP_011508491.1 | |
| XM_017002920.1 | 42 | 6321 | XP_016858409.1 | |
| XM_017002921.1 | 40 | 6273 | XP_016858410.1 | |
| XM_017002922.1 | 41 | 6201 | XP_016858411.1 | |
| XM_017002923.1 | 40 | 6186 | XP_016858412.1 | |
| XM_017002924.1 | 40 | 6186 | XP_016858413.1 | |
| AB002378.1 | 40 | 5487 | ||
| AK302500.1 | 20 | 3379 | ||
| AL832790.1 | 10 | 3813 | ||
| BC057394.1 | 41 | 5926 | AAH57394.1 |
Target Gene Details
Entrez Gene ID: | 149499 |
Gene Name: | leucine rich repeat containing 71 |
Gene Aliases: |
C1orf92 |
Location: |
Chr.1:156920608-156941208 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC71 | XM_017000459.1 | XP_016855948.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890682 | Chr.1:156899922 - 156951428 on Build GRCh38 | Loss |
|
| nsv831659 | Chr.1:156771159 - 156980204 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map