Assay Details
Target Gene Details
Entrez Gene ID: | 126133 |
Gene Name: | aldehyde dehydrogenase 16 family member A1 |
Gene Aliases: |
- |
Location: |
Chr.19:49453216-49471050 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH16A1 | NM_001145396.1 | 4 | 485 | NP_001138868.1 |
| NM_153329.3 | 4 | 485 | NP_699160.2 | |
| XM_011526441.1 | 4 | 303 | XP_011524743.1 | |
| XM_011526442.1 | 4 | 320 | XP_011524744.1 | |
| AK074136.1 | 4 | 475 | BAB84962.1 | |
| AK297101.1 | 4 | 468 | ||
| AK298587.1 | ||||
| AK301246.1 | 2 | 183 | ||
| BC014895.2 | 4 | 445 | AAH14895.2 | |
| DA629821.1 | ||||
| DC364604.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2518 | Chr.19:49452273 - 49465137 on Build GRCh38 | Insertion |
|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv833862 | Chr.19:49448789 - 49570676 on Build GRCh38 | Loss |
|
| nsv953599 | Chr.19:49386444 - 49506743 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map