Assay Details
Target Gene Details
Entrez Gene ID: | 8479 |
Gene Name: | HIRA interacting protein 3 |
Gene Aliases: |
- |
Location: |
Chr.16:29992321-29996096 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIRIP3 | NM_001197323.1 | NP_001184252.1 | ||
| NM_003609.4 | NP_003600.2 | |||
| AJ223349.1 | CAA11273.1 | |||
| AJ223350.1 | CAA11274.1 | |||
| AJ223351.2 | CAA11275.2 | |||
| AK075145.1 | ||||
| AK312504.1 | ||||
| BC000588.2 | AAH00588.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638378 | Chr.16:29973809 - 30006402 on Build GRCh38 | Loss |
|
| esv997840 | Chr.16:29991169 - 30003917 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map