Assay Details
Target Gene Details
Entrez Gene ID: | 23621 |
Gene Name: | beta-secretase 1 |
Gene Aliases: |
ASP2, BACE, HSPC104 |
Location: |
Chr.11:117285686-117316256 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BACE1 | NM_001207048.1 | 8 | 1570 | NP_001193977.1 |
| NM_001207049.1 | 8 | 1495 | NP_001193978.1 | |
| NM_012104.4 | 9 | 2208 | NP_036236.1 | |
| NM_138971.3 | 9 | 2076 | NP_620427.1 | |
| NM_138972.3 | 9 | 2133 | NP_620428.1 | |
| NM_138973.3 | 9 | 2001 | NP_620429.1 | |
| AB032975.2 | 9 | 2190 | BAA86463.2 | |
| AB177847.1 | 8 | 1432 | BAD66825.1 | |
| AF161367.1 | 5 | 1035 | AAF28927.1 | |
| AF190725.1 | 9 | 2200 | AAF04142.1 | |
| AF200193.1 | 9 | 1708 | AAF13715.1 | |
| AF200343.1 | 9 | 1747 | AAF17079.1 | |
| AF201468.1 | 9 | 2201 | AAF18982.1 | |
| AK075049.1 | 9 | 1683 | ||
| AK296535.1 | 8 | 1483 | ||
| BC065492.1 | 9 | 2208 | AAH65492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038815 | Chr.11:117287202 - 117403925 on Build GRCh38 | Gain |
|
| nsv509443 | Chr.11:117255121 - 117293441 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map