Assay Details
Target Gene Details
Entrez Gene ID: | 10260 |
Gene Name: | DENN domain containing 4A |
Gene Aliases: |
IRLB, MYCPBP |
Location: |
Chr.15:65659123-65792293 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND4A | NM_001144823.2 | 29 | 5568 | NP_001138295.1 |
| NM_001320835.1 | 29 | 5571 | NP_001307764.1 | |
| NM_005848.4 | 28 | 5432 | NP_005839.3 | |
| XM_005254120.4 | 28 | 5350 | XP_005254177.1 | |
| XM_005254121.3 | 28 | 5439 | XP_005254178.1 | |
| XM_005254122.4 | 29 | 5553 | XP_005254179.1 | |
| XM_006720371.3 | 29 | 5306 | XP_006720434.1 | |
| XM_011521157.2 | 19 | 3906 | XP_011519459.1 | |
| XM_017021862.1 | 28 | 5291 | XP_016877351.1 | |
| XM_017021863.1 | 27 | 5217 | XP_016877352.1 | |
| XM_017021864.1 | 29 | 5566 | XP_016877353.1 | |
| XM_017021865.1 | 28 | 5420 | XP_016877354.1 | |
| AF534403.1 | 28 | 5400 | AAQ10514.1 | |
| AK091268.1 | 8 | 2149 | ||
| AL832602.2 | 26 | 4958 | CAD89960.1 | |
| AL833317.1 | 29 | 5399 | ||
| AL833384.1 | 25 | 4703 | ||
| BC041706.1 | 28 | 5263 | AAH41706.1 | |
| X63417.1 | CAA45013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833043 | Chr.15:65638528 - 65801425 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map