Assay Details
Target Gene Details
Entrez Gene ID: | 25825 |
Gene Name: | beta-site APP-cleaving enzyme 2 |
Gene Aliases: |
AEPLC, ALP56, ASP1, ASP21, BAE2, CDA13, CEAP1, DRAP |
Location: |
Chr.21:41167801-41282534 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BACE2 | NM_012105.4 | NP_036237.2 | ||
| NM_138991.2 | NP_620476.1 | |||
| NM_138992.2 | NP_620477.1 | |||
| AF050171.1 | AAD45963.1 | |||
| AF117892.1 | AAD45240.1 | |||
| AF178532.1 | AAF29494.1 | |||
| AF188276.1 | AAF35835.1 | |||
| AF188277.1 | AAF35836.1 | |||
| AF200192.1 | AAF13714.1 | |||
| AF200342.1 | AAF17078.1 | |||
| AF204944.1 | AAF26368.1 | |||
| AF212252.1 | AAG41783.1 | |||
| AK075539.1 | BAC11682.1 | |||
| AK292056.1 | ||||
| AY358927.1 | AAQ89286.1 | |||
| BC014453.1 | AAH14453.1 |
Target Gene Details
Entrez Gene ID: | 191585 |
Gene Name: | placenta specific 4 |
Gene Aliases: |
C21orf115, PRED78 |
Location: |
Chr.21:41175231-41185239 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLAC4 | NM_182832.2 | 1 | 8105 | NP_878252.2 |
| AK027868.1 | 1 | 5768 | ||
| AK092431.1 | 2 | 2758 | BAC03888.1 |
More Information
Set Membership: |
|

Genomic Map