Assay Details
Target Gene Details
Entrez Gene ID: | 3249 |
Gene Name: | hepsin |
Gene Aliases: |
TMPRSS1 |
Location: |
Chr.19:35040506-35066573 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPN | NM_002151.2 | NP_002142.1 | ||
| NM_182983.2 | 2 | 669 | NP_892028.1 | |
| XM_005258838.4 | XP_005258895.2 | |||
| XM_006723181.3 | XP_006723244.2 | |||
| XM_017026731.1 | 2 | 342 | XP_016882220.1 | |
| AK315655.1 | ||||
| BC025716.1 | AAH25716.1 | |||
| M18930.1 | AAA36013.1 | |||
| X07732.1 | 2 | 669 | CAA30558.1 |
More Information
Set Membership: |
|

Genomic Map