Assay Details
Target Gene Details
Entrez Gene ID: | 1021 |
Gene Name: | cyclin dependent kinase 6 |
Gene Aliases: |
MCPH12, PLSTIRE |
Location: |
Chr.7:92604921-92836627 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK6 | NM_001145306.1 | 8 | 8492 | NP_001138778.1 |
| NM_001259.6 | 8 | 8387 | NP_001250.1 | |
| XM_006715835.2 | 8 | 8612 | XP_006715898.1 | |
| AB073601.1 | 1 | 392 | ||
| AK055922.1 | 1 | 1360 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819408 | Chr.7:92607938 - 92608315 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113665429] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map