Assay Details
Target Gene Details
Entrez Gene ID: | 23566 |
Gene Name: | lysophosphatidic acid receptor 3 |
Gene Aliases: |
EDG7, Edg-7, GPCR, HOFNH30, LP-A3, LPA3, RP4-678I3 |
Location: |
Chr.1:84813403-84893213 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPAR3 | NM_012152.2 | 3 | 1245 | NP_036284.1 |
| AF127138.1 | 2 | 1071 | AAD56311.1 | |
| AF186380.1 | AAF00530.1 | |||
| AF236117.1 | AAF91291.1 | |||
| AK308283.1 | 3 | 1248 | ||
| AY322547.1 | AAP84360.1 | |||
| BC126268.1 | 2 | 1051 | ||
| BC126270.1 | 2 | 1051 | ||
| BU633960.1 | 1 | 494 |
More Information
Set Membership: |
|

Genomic Map