Assay Details
Target Gene Details
Entrez Gene ID: | 10730 |
Gene Name: | YME1 like 1 ATPase |
Gene Aliases: |
FTSH, MEG4, PAMP, YME1L |
Location: |
Chr.10:27110111-27154802 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YME1L1 | NM_001253866.1 | 4 | 603 | NP_001240795.1 |
| NM_014263.3 | 5 | 702 | NP_055078.1 | |
| NM_139312.2 | 6 | 873 | NP_647473.1 | |
| XM_011519300.2 | 5 | 1156 | XP_011517602.1 | |
| AF070656.1 | AAD20962.1 | |||
| AF151782.1 | 6 | 717 | AAK57555.1 | |
| AJ132637.1 | 5 | 675 | CAB51858.1 | |
| AK291292.1 | 5 | 631 | ||
| AK297973.1 | 4 | 567 | ||
| AK309708.1 | 1 | 149 | ||
| AK314372.1 | 5 | 630 | ||
| AY358484.1 | 5 | 628 | AAQ88848.1 | |
| BC007795.2 | 6 | 809 | AAH07795.1 | |
| BC023507.2 | 5 | 644 | AAH23507.1 | |
| BC024032.2 | 5 | 649 | AAH24032.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2664616 | Chr.10:26927640 - 27310597 on Build GRCh38 | Deletion |
|
| esv3622707 | Chr.10:27071518 - 27136388 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map