Assay Details
Target Gene Details
Entrez Gene ID: | 861 |
Gene Name: | runt related transcription factor 1 |
Gene Aliases: |
AML1, AML1-EVI-1, AMLCR1, CBF2alpha, CBFA2, EVI-1, PEBP2aB, PEBP2alpha |
Location: |
Chr.21:34787801-35049334 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RUNX1 | NM_001001890.2 | 6 | 5589 | NP_001001890.1 |
| NM_001754.4 | 9 | 4282 | NP_001745.2 | |
| XM_005261068.3 | 7 | 4545 | XP_005261125.1 | |
| XM_005261069.4 | 8 | 4126 | XP_005261126.1 | |
| XM_011529766.2 | 9 | 4393 | XP_011528068.1 | |
| XM_011529767.2 | 8 | 4279 | XP_011528069.1 | |
| XM_011529768.2 | 7 | 4087 | XP_011528070.1 | |
| XM_017028487.1 | 7 | 4087 | XP_016883976.1 | |
| D43968.1 | 6 | 5589 | BAA07903.1 | |
| D43969.1 | 8 | 4536 | BAA07904.1 | |
| X90976.1 | 1 | 2694 | CAA62464.1 |
More Information
Set Membership: |
|

Genomic Map