Assay Details
Target Gene Details
Entrez Gene ID: | 10436 |
Gene Name: | EMG1, N1-specific pseudouridine methyltransferase |
Gene Aliases: |
C2F, Grcc2f, NEP1 |
Location: |
Chr.12:6970781-6997428 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EMG1 | NR_135131.1 | |||
| AK298745.1 |
Target Gene Details
Entrez Gene ID: | 10162 |
Gene Name: | lysophosphatidylcholine acyltransferase 3 |
Gene Aliases: |
C3F, LPCAT, LPLAT 5, LPSAT, MBOAT5, OACT5, nessy |
Location: |
Chr.12:6976185-7018538 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPCAT3 | NM_005768.5 | NP_005759.4 | ||
| AK058063.1 | ||||
| AK096775.1 | ||||
| AK296145.1 | ||||
| AK315538.1 | ||||
| BC000664.2 | AAH00664.2 | |||
| BC065194.1 | AAH65194.1 | |||
| BT007000.1 | AAP35646.1 | |||
| BX647983.1 | ||||
| BX648009.1 | ||||
| U72515.1 | AAC51640.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047373 | Chr.12:6902891 - 7055754 on Build GRCh38 | Gain |
|
| nsv509453 | Chr.12:6896530 - 7007852 on Build GRCh38 | Insertion |
|
| nsv1035811 | Chr.12:6876316 - 6994911 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map