Assay Details
Target Gene Details
Entrez Gene ID: | 1514 |
Gene Name: | cathepsin L |
Gene Aliases: |
CATL, CTSL1, MEP |
Location: |
Chr.9:87725437-87731469 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTSL | NM_001257971.1 | NP_001244900.1 | ||
| NM_001257972.1 | NP_001244901.1 | |||
| NM_001257973.1 | NP_001244902.1 | |||
| NM_001912.4 | NP_001903.1 | |||
| NM_145918.2 | NP_666023.1 | |||
| XM_005251716.3 | XP_005251773.1 | |||
| XM_011518263.1 | XP_011516565.1 | |||
| XM_017014293.1 | XP_016869782.1 | |||
| AF217997.1 | AAG17239.1 | |||
| AK055599.1 | ||||
| AK075100.1 | ||||
| AL832167.1 | ||||
| BC012612.1 | AAH12612.1 | |||
| BC142983.1 | ||||
| BM680778.1 | ||||
| BX537395.1 | CAD97637.1 | |||
| BX647101.1 | ||||
| BX647102.1 | ||||
| BX647413.1 | ||||
| BX647434.1 | ||||
| BX647435.1 | ||||
| BX648848.1 | ||||
| BX648849.1 | ||||
| BX649140.1 | ||||
| CR457053.1 | CAG33334.1 | |||
| X12451.1 | CAA30981.1 | |||
| Y18462.1 | CAA77180.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759703 | Chr.9:87403812 - 87810065 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map