Assay Details
Target Gene Details
Entrez Gene ID: | 6603 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2 |
Gene Aliases: |
BAF60B, CRACD2, PRO2451, Rsc6p |
Location: |
Chr.17:63832081-63842991 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMARCD2 | NM_001098426.1 | 10 | 1571 | NP_001091896.1 |
| XM_005257604.2 | 10 | 1118 | XP_005257661.2 | |
| XM_017024967.1 | 10 | 2310 | XP_016880456.1 | |
| AK025917.1 | 2 | 135 | ||
| AK300939.1 | 10 | 1355 | ||
| AK303951.1 | 10 | 1157 | ||
| AK307944.1 | 10 | 1124 | ||
| BC136322.1 | 11 | 1440 | ||
| BC142963.1 | 10 | 1520 | ||
| U66618.1 | 10 | 1514 | AAC50696.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833510 | Chr.17:63767293 - 63927790 on Build GRCh38 | Loss |
|
| nsv517272 | Chr.17:63814307 - 64020132 on Build GRCh38 | Gain+Loss |
|
| nsv510720 | Chr.17:63809351 - 63908780 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map